Learn More
Beckman Coulter Racks
Catalog No. NC9730137
Encompass
The Beckman Coulter Microplate Racks are designed to securely hold microplates during incubation and washing steps. They are made from durable polypropylene and are autoclavable. The racks have alphanumeric grid reference for easy plate location. They come in different capacities including 1, 2, 3 or 4 plates to fit different workflow needs. The racks have a small footprint to conserve benchtop space. Overall, these are high-quality racks to securely hold microplates during various lab procedures.
Catalog No. NC9730137 Supplier Beckman Coulter Supplier No. 974627
The Beckman Coulter Microplate Racks are designed to securely hold microplates during incubation and washing steps. They are made from durable polypropylene and are autoclavable. The racks have alphanumeric grid reference for easy plate location. They come in different capacities including 1, 2, 3 or 4 plates to fit different workflow needs. The racks have a small footprint to conserve benchtop space. Overall, these are high-quality racks to securely hold microplates during various lab procedures.