Learn More
Advantec MFS, Inc Vacuum Filtration Manifold; PVC, 6-Branch with 2-Way Valves
Supplier:  Advantec MFS, Inc 313600
Encompass_Preferred
Product features: With Standard Cup, 4.75" Distance Between Stations, 34.5" L x 4.75" W x 7" H. PVC Manifolds have branches with two-way PTFE valves for independent control of vacuum. Hose barb accepts 1⁄4" ID tubing. Branches accept no. 8 stoppers. Sanitize with ethanol or formaldehyde; do not autoclave. Maximum temperature is 140°F (60°C). Distance between stations is 4.75" (12 cm). Significantly reduce process time compared to gravity methods Two- and three-way valves feature PTFE stopcocks for smooth control. Valves provide independent control at each branch for vacuum and venting. Connect three or six filter holders to one vacuum source
Catalog No. NC0463926