Learn More
USA Scientific Inc 6-place (3 x 2) upright freezer rack for 4 3/4 inch H boxes
Catalog No. NC1734655
Encompass
6-place (3 long x 2 high) upright freezer rack for 4 3/4 in H boxes, 18 1/4 in L x 5 7/8 in W x 9 7/8 in H , stainless steel
Catalog No. NC1734655 Supplier USA Scientific Inc Supplier No. 26239006
6-place (3 long x 2 high) upright freezer rack for 4 3/4 in H boxes, 18 1/4 in L x 5 7/8 in W x 9 7/8 in H , stainless steel