Learn More
Research Products International Corp 6"" Platform Head w/o Insert for Vortex-Genie
SDP
Catalog No. 501978251
Encompass
The 6 Inch (152mm) Platform can be used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse in conjunction with either the Microtube Foam Insert, Microplate Foam Insert or Two-Tier Microplate Foam Insert.
Catalog No. 50-197-8251 Supplier Research Products International Corp Supplier No. 1466005
The 6 Inch (152mm) Platform can be used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse in conjunction with either the Microtube Foam Insert, Microplate Foam Insert or Two-Tier Microplate Foam Insert.