Learn More
Ampco 6-Point Standard Impact Socket with 1/2 in. Drive
SDP
Catalog No. 19173966 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Chuck Size:
1 in.
1/2 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chuck Size
19-173-966 1 in.
19-173-967 1/2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-173-966 Supplier Ampco Supplier No. I1/2D1
Only null left
Add to Cart
Add to Cart
Non-sparking, non-magnetic, corrosion resistant standard impact socket. Made in the USA.
  • Standard 6-point impact socket.
  • Made in the USA.
Specifications
Chuck Size 1 in.
Product Type Standard Impact Socket
Drive Size 1/2 in.
Length (English) 1 -3/16 in.
Includes One 1/2 in. Socket Size Socket