Learn More
Waters Corp Waters Rotor, 6 Port, 2 Position, LC Injection Valve
Catalog No. 50792698 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rotor is designed for use in HPLC systems as a replacement valve rotor. It is essential for preventative maintenance or when issues like injection problems, inaccuracy, contamination, or carryover are observed, ensuring optimal instrument performance.
  • Used for preventative maintenance
  • Addresses injection problems, inaccuracy, contamination, or carryover
  • Features 6 ports
  • Has 2 positions
  • Suitable for LC injection valves
  • Designed for HPLC systems
  • Compatible with DC6WK valve
Catalog No. 50-792-698 Supplier Waters Corp Supplier No. 700002210
This rotor is designed for use in HPLC systems as a replacement valve rotor. It is essential for preventative maintenance or when issues like injection problems, inaccuracy, contamination, or carryover are observed, ensuring optimal instrument performance.
  • Used for preventative maintenance
  • Addresses injection problems, inaccuracy, contamination, or carryover
  • Features 6 ports
  • Has 2 positions
  • Suitable for LC injection valves
  • Designed for HPLC systems
  • Compatible with DC6WK valve