Learn More
Thermo Scientific 6 Port Standard Seal
Catalog No. NC9625882 Shop All Thermo Scientific Products
Encompass
Multiport seal for laboratory and analytical equipment enables simultaneous connection of up to 6 sample or reagent lines into sealed system for leak-free transfer of liquids or gases
Catalog No. NC9625882 Supplier Thermo Scientific Supplier No. CH953197
Multiport seal for laboratory and analytical equipment enables simultaneous connection of up to 6 sample or reagent lines into sealed system for leak-free transfer of liquids or gases