Learn More
DSI LLC 6 POS VLV,1/16,.040,2S0,200C,400PSI,N6
Catalog No. 502116914
Encompass_Preferred
6 POS VLV,1/16,.040,2S0,200C,400PSI,N6
Catalog No. 50-211-6914 Supplier DSI LLC Supplier No. 2LCSD6MWE
6 POS VLV,1/16,.040,2S0,200C,400PSI,N6