Learn More
Analytical Sales & Services 6 Position, 7 Port Low Pressure Selector Valve with Fittingless Tubing Connection
Catalog No. 5000806655
Encompass
Stainless Steel 6 Position, 7 Port Low Pressure Analytical Selector Valve with Fittingless Tubing Connection. 125psi (9 bar)
Qty/Pkg: EACH
Catalog No. 50-008-06655 Supplier Analytical Sales & Services Supplier No. VXL554670
Stainless Steel 6 Position, 7 Port Low Pressure Analytical Selector Valve with Fittingless Tubing Connection. 125psi (9 bar)
Qty/Pkg: EACH