Learn More
LUNA NANOTECH INC 6-Position Midi Magnetic Separation Rack For 15 ML Tubes
Catalog No. 502566965
Encompass_Preferred
MIDI Magnetic Separation Rack for 15 mL Tubes
MIDI format magnetic racks for medium scale separation of magnetic beads in 15 mL centrifuge tubes. These racks can be used with our PuroMAG MIDI kits as well as various magnetic beads offered by Luna Nanotech.
Medium 15 mL tube volume makes the rack optimal when performing mdium scale magnetic separation reactions.
- Fits 15 mL centrifuge tubes.
- Available in 8-position format.
Catalog No. 50-256-6965 Supplier LUNA NANOTECH INC Supplier No. MGRD008
MIDI Magnetic Separation Rack for 15 mL Tubes
MIDI format magnetic racks for medium scale separation of magnetic beads in 15 mL centrifuge tubes. These racks can be used with our PuroMAG MIDI kits as well as various magnetic beads offered by Luna Nanotech.
Medium 15 mL tube volume makes the rack optimal when performing mdium scale magnetic separation reactions.
- Fits 15 mL centrifuge tubes.
- Available in 8-position format.