Learn More
JDM LAB SOLUTIONS LLC 6 POSITION-MULTI MAG STIRRER
Catalog No. NC1866748
Encompass
6 Position Magnetic Stirrer. The front control panel has the ability to detach from the main unit. This unique feature allows the main stirring base to be placed & operated in a water bath.
Catalog No. NC1866748 Supplier JDM LAB SOLUTIONS LLC Supplier No. MS6BSUA
6 Position Magnetic Stirrer. The front control panel has the ability to detach from the main unit. This unique feature allows the main stirring base to be placed & operated in a water bath.