Learn More
Electron Microscopy Sciences Foam Over Cotton Chemical Resistant Applicator Polypropylene Shaft, 6" OAL, Tip
SDP
Catalog No. 50949197
Encompass_Preferred
Static-dissipative swabs attract dirt and dust for easy cleaning, while reducing and dissipating static electricity. Antistatic properties will not decay over time. Swabs meet all industry standards and requirements, including ASTM-D257 and Federal test method 4046, standard #101C. 6" polypropylene shaft. Tip: 3⁄8"Dia x 15⁄16"L
OAL: 6"
Catalog No. 50-949-197 Supplier Electron Microscopy Sciences Supplier No. 72318
Static-dissipative swabs attract dirt and dust for easy cleaning, while reducing and dissipating static electricity. Antistatic properties will not decay over time. Swabs meet all industry standards and requirements, including ASTM-D257 and Federal test method 4046, standard #101C. 6" polypropylene shaft. Tip: 3⁄8"Dia x 15⁄16"L
OAL: 6"