Learn More
Cole-Parmer Bain Marie HDPE Container, 6 quart; 12/Pk
Catalog No. NC1761829 Shop All Cole-Parmer Products
Encompass_Preferred
Containers are made of FDA-compliant HDPE and can be frozen, boiled, or chemically sterilized. Snap-tight, positive-lock lids are sold separately. NSF certified.
Catalog No. NC1761829 Supplier Cole-Parmer Supplier No. 6353004
Containers are made of FDA-compliant HDPE and can be frozen, boiled, or chemically sterilized. Snap-tight, positive-lock lids are sold separately. NSF certified.