Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Quinolinecarbaldehyde, 97%, Thermo Scientific™

Catalog No. CC04504DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC04504DE 5 g Amber Glass Bottle
CC04504CB 250 mg Amber Glass Bottle
CC04504DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC04504DE Supplier Thermo Scientific Chemicals Supplier No. CC04504DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4113-04-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
InChI Key VUAOIXANWIFYCU-UHFFFAOYSA-N
Synonym quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
PubChem CID 765653

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material 6-Quinolinecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.