Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Quinoxalinamine, 97%, Thermo Scientific™

Catalog No. CC11714DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC11714DA 1 g Amber Glass Bottle
CC11714EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC11714DA Supplier Thermo Scientific Chemicals Supplier No. CC11714DA
Add to Cart

Chemical Identifiers

CAS 6298-37-9
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00462821
InChI Key MSGRFBKVMUKEGZ-UHFFFAOYSA-N
Synonym 6-aminoquinoxaline, 6-quinoxalinamine, 6-quinoxalinylamine, quinoxalin-6-ylamine, quinoxaline-6-ylamine, 6-chinoxalinamin, 6-amino-quinoxaline, pubchem15453, quinoxalin, 6-amino
PubChem CID 237859
IUPAC Name quinoxalin-6-amine
SMILES NC1=CC=C2N=CC=NC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material 6-Quinoxalinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.