Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Ring NOGE (Novolac glycidyl ether) Mixture of Isomers, Analytical Standard, ≥80.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111003394 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3394 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3394 Supplier MilliporeSigma Supelco Supplier No. 3097750MGF
Add to Cart
Add to Cart

Chain-like or Branched

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Synonym NOGE 6-Ring (Novolac glycidyl ether) mixture of isomers; Novolac glycidyl ether 6-Ring
Quantity 50 mg
Percent Purity ≥80.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.