Learn More
AdipoGen 6-ROX
SDP
Supplier:  AdipoGen CDXC0084M125
Encompass_Preferred
Chemical. CAS 194785-18-7. Formula C33H30N2O5. MW 534.6. Synthetic. This pure-6 isomer is a derivative of carboxy-X-rhodamine, widely used for oligonucleotide labeling and automated DNA sequencing applications.
Catalog No. 50-256-1485