Learn More
Medchemexpress LLC 6-ROX (6-Carboxy-X-rhodamine) | 194785-18-7 | 98.4% | 100 MG
SDP
Catalog No. 5000409774
Encompass_Preferred
6-ROX is a selective fluorescent probe and a potential inhibitor of COX-2. It binds to the active site of COX-2, preventing the conversion of arachidonic acid into prostaglandins. This compound is frequently utilized in optical imaging for tumors and inflammation, aiding in the detection of diseased tissues that highly express COX-2.
  • Acts as a selective fluorescent probe
  • Potential inhibitor of COX-2
  • Binds to the active site of COX-2
  • Prevents conversion of arachidonic acid into prostaglandins
  • Utilized in optical imaging for tumors and inflammation
  • Aids in detection of diseased tissues with high COX-2 expression
Catalog No. 50-004-09774 Supplier Medchemexpress LLC Supplier No. HYD0053100MG
6-ROX is a selective fluorescent probe and a potential inhibitor of COX-2. It binds to the active site of COX-2, preventing the conversion of arachidonic acid into prostaglandins. This compound is frequently utilized in optical imaging for tumors and inflammation, aiding in the detection of diseased tissues that highly express COX-2.
  • Acts as a selective fluorescent probe
  • Potential inhibitor of COX-2
  • Binds to the active site of COX-2
  • Prevents conversion of arachidonic acid into prostaglandins
  • Utilized in optical imaging for tumors and inflammation
  • Aids in detection of diseased tissues with high COX-2 expression