Learn More
Biotium 6-ROX, FREE ACID
Catalog No. 501964802
Encompass_Preferred
6-Carboxy-X-rhodamine is a single isomer, long wavelength rhodamine dye.Properties: Ex/ Em (MeOH) = 568/595 nm; Dark red solid soluble in DMSO, DMF or MeOH; Store at -20°C and protect from light; C 33 H 30 N 2 O 5; MW: 534.6
Catalog No. 50-196-4802 Supplier Biotium Supplier No. 90013
6-Carboxy-X-rhodamine is a single isomer, long wavelength rhodamine dye.Properties: Ex/ Em (MeOH) = 568/595 nm; Dark red solid soluble in DMSO, DMF or MeOH; Store at -20°C and protect from light; C 33 H 30 N 2 O 5; MW: 534.6