Learn More
Aat Bioquest 6-ROX, SE [6-Carboxy-X-rhodamine, succinimidyl ester] *CAS#: 216699-36-4*
SDP
Catalog No. 501952873
Encompass_Preferred
6-ROX, SE is the other purified single isomer of 5(6)-ROX, SE. 6-ROX is predominately used for labeling nucleotides and sequencing nucleic acids. Compared to other rhodamines, ROX is very unstable.
Catalog No. 50-195-2873 Supplier Aat Bioquest Supplier No. 392
6-ROX, SE is the other purified single isomer of 5(6)-ROX, SE. 6-ROX is predominately used for labeling nucleotides and sequencing nucleic acids. Compared to other rhodamines, ROX is very unstable.