Learn More
Ace Glass, Inc. 6 SECTION SNYDER COLUMN 6 SECTION SNYDER COLUMN
SDP
Catalog No. 50973277
Encompass_Preferred
6-section Snyder column for use in distillation applications. Column is floating ball type with 24/40 joints. Each section is approximately 51mm long.
Catalog No. 50-973-277 Supplier Ace Glass, Inc. Supplier No. 657504
6-section Snyder column for use in distillation applications. Column is floating ball type with 24/40 joints. Each section is approximately 51mm long.