Learn More
Cayman Chemical 6-SIalylactosesodIum salt 10mg
SDP
Catalog No. 502967921
Encompass_Preferred
A major milk oligosaccharides that avidly bind several viral strains, including strains of influenza, HIV-1, reovirus, and polyomavirus; used to differentiate and characterize the binding domains of viruses that recognize N-acetylneuraminic acid-capped cell surface receptors; also used as an analytical reference standard for quantification in samples such as milk or colostrum
Catalog No. 50-296-7921 Supplier Cayman Chemical Supplier No. 1786610 MG
A major milk oligosaccharides that avidly bind several viral strains, including strains of influenza, HIV-1, reovirus, and polyomavirus; used to differentiate and characterize the binding domains of viruses that recognize N-acetylneuraminic acid-capped cell surface receptors; also used as an analytical reference standard for quantification in samples such as milk or colostrum