Learn More
Cayman Chemical 6'-Sialyllactose (sodium salt), Purity: ≥98%, CAS: 157574-76-0, 50 mg
SDP
Catalog No. 502965880
Encompass_Preferred
A major milk oligosaccharides that avidly bind several viral strains, including strains of influenza, HIV-1, reovirus, and polyomavirus; used to differentiate and characterize the binding domains of viruses that recognize N-acetylneuraminic acid-capped cell surface receptors; also used as an analytical reference standard for quantification in samples such as milk or colostrum
Catalog No. 50-296-5880 Supplier Cayman Chemical Supplier No. 1786650 MG
A major milk oligosaccharides that avidly bind several viral strains, including strains of influenza, HIV-1, reovirus, and polyomavirus; used to differentiate and characterize the binding domains of viruses that recognize N-acetylneuraminic acid-capped cell surface receptors; also used as an analytical reference standard for quantification in samples such as milk or colostrum