Learn More
Sigma Aldrich Fine Chemicals Biosciences 6 SigMatrix Serum diluent
Catalog No. 502932463
Encompass
This reagent is a 6% recombinant HSA (Human Serum Albumin) in PBS SOLUTIONon pH 7.4 that can be used as a blank or matrix/diluent for proteinaceous analytes in LC-MS/MS calibrators controls or samples.The 6% protein content design is meant to mimic endogenous protein concentration in human serum samples.Beneficially as the HSA source is recombinant the matrix should not contain any measurable contaminants typically found in even stripped serum.
Catalog No. 50-293-2463 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D532250ML
This reagent is a 6% recombinant HSA (Human Serum Albumin) in PBS SOLUTIONon pH 7.4 that can be used as a blank or matrix/diluent for proteinaceous analytes in LC-MS/MS calibrators controls or samples.The 6% protein content design is meant to mimic endogenous protein concentration in human serum samples.Beneficially as the HSA source is recombinant the matrix should not contain any measurable contaminants typically found in even stripped serum.