Learn More
AdipoGen 6-TAMRA
SDP
Supplier:  AdipoGen CDXC0056M025
Encompass_Preferred
Chemical. CAS 91809-67-5. Formula C25H22N2O5. MW 430.45. Synthetic. Free acid form of 6-carboxytetramethylrhodamine single isomer. The corresponding succinimidyl ester is a single isomer fluorophore most commonly used in automated DNA sequencing.
Catalog No. 50-256-1505