Learn More
Aat Bioquest 6-TAMRA cadaverine
SDP
Catalog No. 501952846
Encompass_Preferred
6-TAMRA cadaverine is a building block for developing fluoresceinated bioconjugates. It is also a fluorescent transglutaminase substrate to label proteins by transaminidation.
Catalog No. 50-195-2846 Supplier Aat Bioquest Supplier No. 357
6-TAMRA cadaverine is a building block for developing fluoresceinated bioconjugates. It is also a fluorescent transglutaminase substrate to label proteins by transaminidation.