Learn More
AdipoGen 6-TAMRA N-succinimidyl ester
SDP
Supplier:  AdipoGen CDXC0058M025
Encompass_Preferred
Chemical. CAS 150810-69-8. Formula C29H25N3O7. MW 527.52. Synthetic. Amine-reactive form of 6-carboxytetramethylrhodamine single isomer used for labeling of peptides and aminoglycoside antibiotics. It is also most commonly used in automated DNA sequencing.
Catalog No. 50-256-1499