Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-tert-Butylquinoline 98.0+%, TCI America™
SDP

Catalog No. B29851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B29851G Supplier TCI America Supplier No. B29851G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68141-13-9
Molecular Formula C13H15N
Molecular Weight (g/mol) 185.27
MDL Number MFCD02683023
InChI Key JHAWWJQGHKGXHA-UHFFFAOYSA-N
PubChem CID 109638
IUPAC Name 6-tert-butylquinoline
SMILES CC(C)(C)C1=CC=C2N=CC=CC2=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 185.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 6-tert-Butylquinoline
EINECSNumber (5)-0796
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.