Learn More
AdipoGen 6-TET dipivaloate
SDP
Catalog No. 502561433
Encompass_Preferred
Chemical. CAS 314734-87-7. Formula C31H24Cl4O9. MW 751.2274. Synthetic. 6-TET is an amino-conjugatable fluorescent probe that is widely used in nucleic acid sequencing and related research. It can also be used to label peptides and oligonucleotides.
Catalog No. 50-256-1433 Supplier AdipoGen Supplier No. CDXC0186M500
Chemical. CAS 314734-87-7. Formula C31H24Cl4O9. MW 751.2274. Synthetic. 6-TET is an amino-conjugatable fluorescent probe that is widely used in nucleic acid sequencing and related research. It can also be used to label peptides and oligonucleotides.