Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-thien-2-ylnicotinic acid, Thermo Scientific™

Catalog No. CC54801CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC54801CB 250 mg
CC54801DA 1 g
CC54801DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC54801CB Supplier Thermo Scientific Chemicals Supplier No. CC54801CB
Add to Cart

Chemical Identifiers

CAS 179408-54-9
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.23
MDL Number MFCD06410384
InChI Key QYGFXNUFKLRCRB-UHFFFAOYSA-N
Synonym 6-thien-2-ylnicotinic acid, 6-thiophen-2-yl nicotinic acid, 6-2-thienyl nicotinic acid, 6-thiophen-2-yl pyridine-3-carboxylic acid, 6-2-thienyl pyridine-3-carboxylic acid, acmc-209efk, 6-thien-2ylnicotinic acid, 6-thiophen-2-yl-nicotinic acid, 6-thiophen-2-ylnicotinic acid, 2-thien-2-yl-5-pyridinecarboxylic acid
PubChem CID 15411144
SMILES OC(=O)C1=CN=C(C=C1)C1=CC=CS1

Specifications

Assay Percent Range 97%
Quantity 250 mg
IUPAC Name 6-thiophen-2-ylpyridine-3-carboxylic acid
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 6-Thien-2-ylnicotinic acid
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.