Learn More
TARGETMOL CHEMICALS INC 6-THIO-2-DEOXYGUANOSINE 25MG
SDP
Catalog No. 502579294
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 6-Thio-2'-Deoxyguanosine ((beta)-TGdR) is a nucleoside analog and telomerase substrate. purity: 99%
Catalog No. 50-257-9294 Supplier TARGETMOL CHEMICALS INC Supplier No. T674625MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 6-Thio-2'-Deoxyguanosine ((beta)-TGdR) is a nucleoside analog and telomerase substrate. purity: 99%