Learn More
Medchemexpress LLC 6-Thio-2'-deoxyguanosine | 789-61-7 | 99.7% | 283.31 | 10 MG
SDP
Catalog No. 5000365363
Encompass_Preferred
6-Thio-2'-Deoxyguanosine is a nucleoside analogue that can be incorporated into de novo-synthesized telomeres by telomerase. This compound has shown rapid cell death in various cancer cell lines with IC50 values between 0.7-2.9 μM. In A549 lung cancer cell-based mouse xenograft studies, it decreased tumor growth rate, outperforming 6-thioguanine treatment, and increased telomere dysfunction in tumor cells in vivo. It is for research use only.
  • Nucleoside analogue
  • Incorporated into de novo-synthesized telomeres by telomerase
  • Induces rapid cell death in various cancer cell lines
  • Decreases tumor growth rate in mouse xenograft studies
  • Increases telomere dysfunction in tumor cells in vivo
Catalog No. 50-003-65363 Supplier Medchemexpress LLC Supplier No. 10MGHY18762
6-Thio-2'-Deoxyguanosine is a nucleoside analogue that can be incorporated into de novo-synthesized telomeres by telomerase. This compound has shown rapid cell death in various cancer cell lines with IC50 values between 0.7-2.9 μM. In A549 lung cancer cell-based mouse xenograft studies, it decreased tumor growth rate, outperforming 6-thioguanine treatment, and increased telomere dysfunction in tumor cells in vivo. It is for research use only.
  • Nucleoside analogue
  • Incorporated into de novo-synthesized telomeres by telomerase
  • Induces rapid cell death in various cancer cell lines
  • Decreases tumor growth rate in mouse xenograft studies
  • Increases telomere dysfunction in tumor cells in vivo