Learn More
Medchemexpress LLC 6-Thio-2'-Deoxyguanosine | 789-61-7 | 99.7% | 283.31 | 25 MG
SDP
Catalog No. 5000386030
Encompass_Preferred
6-Thio-2'-Deoxyguanosine is a nucleoside analogue that can be incorporated into de novo-synthesized telomeres by telomerase. This product is for research use only and not for sale to patients.
  • Treatment results in rapid cell death for the vast majority of cancer cell lines tested, with IC50s between 0.7-2.9 μM.
  • In A549 lung cancer cell-based mouse xenograft studies, it causes a decrease of the tumor growth rate, superior to that observed with 6-thioguanine treatment.
  • It increases telomere dysfunction in tumor cells in vivo.
Catalog No. 50-003-86030 Supplier Medchemexpress LLC Supplier No. 25MGHY18762
6-Thio-2'-Deoxyguanosine is a nucleoside analogue that can be incorporated into de novo-synthesized telomeres by telomerase. This product is for research use only and not for sale to patients.
  • Treatment results in rapid cell death for the vast majority of cancer cell lines tested, with IC50s between 0.7-2.9 μM.
  • In A549 lung cancer cell-based mouse xenograft studies, it causes a decrease of the tumor growth rate, superior to that observed with 6-thioguanine treatment.
  • It increases telomere dysfunction in tumor cells in vivo.