Learn More
Cayman Chemical 6-thIo-2-DeoxygunosIn 5mg
SDP
Catalog No. 502983295
Encompass_Preferred
A purine nucleoside analog; reduces infection of Calu-3 cells by SARS-CoV-2 (EC50 = 0.04 µM) but is also toxic to Calu-3 cells (CC50 = 1.14 µM); decreases tumor volume and increases the number of tumor cells with telomere dysfunction-induced foci in an A549 NSCLC mouse xenograft model at 2 mg/kg; increases the number of PD-1+ CD8+ T cells, as well as induces tumor regression when administered in combination with anti-PD-L1 in MC-38 murine colon carcinoma models
Catalog No. 50-298-3295 Supplier Cayman Chemical Supplier No. 191545 MG
Add to Cart
Add to Cart
A purine nucleoside analog; reduces infection of Calu-3 cells by SARS-CoV-2 (EC50 = 0.04 µM) but is also toxic to Calu-3 cells (CC50 = 1.14 µM); decreases tumor volume and increases the number of tumor cells with telomere dysfunction-induced foci in an A549 NSCLC mouse xenograft model at 2 mg/kg; increases the number of PD-1+ CD8+ T cells, as well as induces tumor regression when administered in combination with anti-PD-L1 in MC-38 murine colon carcinoma models