Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Thioguanine 95.0+%, TCI America™
SDP

Catalog No. T02125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02125G 5 g
T02121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T02125G Supplier TCI America Supplier No. T02125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00233553
InChI Key WYWHKKSPHMUBEB-UHFFFAOYSA-N
Synonym 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
PubChem CID 2723601
ChEBI CHEBI:9555
IUPAC Name 2-amino-6,7-dihydro-3H-purine-6-thione
SMILES NC1=NC(=S)C2=C(N1)N=CN2

Specifications

Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 167.19
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Thioguanine
RTECSNumber UP0740000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.