Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Thioguanine, Tocris Bioscience™
SDP

Catalog No. 406150
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-615-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-615-0 Supplier R&D Systems Supplier No. 4061/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anticancer and immunosuppressive agent

Anticancer and immunosuppressive agent often used to treat immune disorders and leukemia. Displays cytotoxic and antineoplastic properties; disrupts cytosine methylation by DNA methyltransferases after incorporation into DNA. Selectively kills BRCA2-defective tumors in a xenograft model. Also facilitates proteasome-mediated degradation of DNA (cytosine-5)-methyltransferase 1 (DNMT1). Also inhibits SARS-CoV papain-like protease (PLpro; IC50 = 5 μM for PLpro deubiquitination activity).

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00233553
InChI Key WYWHKKSPHMUBEB-UHFFFAOYSA-N
Synonym 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
PubChem CID 2723601
ChEBI CHEBI:9555
IUPAC Name 2-amino-6,7-dihydro-3H-purine-6-thione
SMILES NC1=NC(=S)C2=C(N1)N=CN2

Specifications

Quantity 50 mg
Formula Weight 167.19
Percent Purity >99%
Chemical Name or Material 6-Thioguanine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.