Learn More
Cayman Chemical 6-Thioguanosine, Purity: ≥98%, CAS: 85-31-4, 500 mg
SDP
Catalog No. 502912215
Encompass_Preferred
A thiopurine nucleoside; preincubation decreases uridine and methionine incorporation into ribosomal RNA in Novikoff-Hepatoma cells at 10 and 100 µM, respectively; inhibits the germination of B. anthracis spores (Ki = 98 µM) and prevents spore-induced cytotoxicity in RAW 264.7 macrophages in a concentration-dependent manner; is active against E. coli (IC50 = 2 µM); reduces the proliferation of MOLT-4 leukemia cells (IC50 = 0.19 µM)
Catalog No. 50-291-2215 Supplier Cayman Chemical Supplier No. 36539500 MG
A thiopurine nucleoside; preincubation decreases uridine and methionine incorporation into ribosomal RNA in Novikoff-Hepatoma cells at 10 and 100 µM, respectively; inhibits the germination of B. anthracis spores (Ki = 98 µM) and prevents spore-induced cytotoxicity in RAW 264.7 macrophages in a concentration-dependent manner; is active against E. coli (IC50 = 2 µM); reduces the proliferation of MOLT-4 leukemia cells (IC50 = 0.19 µM)