Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-thioinosine phosphate (thioinosinic acid) | 53-83-8 | MFCD18252517 | 90.0% | 364.27 g/mol | C10H13N4O7PS | 1 MG
SDP

Catalog No. 5000285107
Encompass_Preferred

6-Thioinosine Phosphate (thioinosinic acid) is the phosphorylated metabolite of 6-thioinosine and an intermediate metabolite of the immunosuppressive purine analog azathioprine. Supplied as a white to off-white solid for research use only, it is used in biochemical and pharmacological studies of nucleotide metabolism and drug metabolism.

  • Intermediate metabolite of azathioprine and thiopurine metabolism.
  • Phosphorylated nucleoside (monophosphate) compatible with biochemical assays.
  • Used to study nucleotide metabolism and azathioprine pharmacodynamics.
  • White to off-white solid appearance suitable for analytical use.
  • Molecular formula C10H13N4O7PS; molecular weight 364.27 g/mol.
  • CAS number 53-83-8 for unambiguous identification.
  • Purity 90.0% as supplied.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-85107 Supplier Medchemexpress LLC Supplier No. HYW3544791MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Thioinosine Phosphate (thioinosinic acid) is the phosphorylated metabolite of 6-thioinosine and an intermediate metabolite of the immunosuppressive purine analog azathioprine. Supplied as a white to off-white solid for research use only, it is used in biochemical and pharmacological studies of nucleotide metabolism and drug metabolism.

  • Intermediate metabolite of azathioprine and thiopurine metabolism.
  • Phosphorylated nucleoside (monophosphate) compatible with biochemical assays.
  • Used to study nucleotide metabolism and azathioprine pharmacodynamics.
  • White to off-white solid appearance suitable for analytical use.
  • Molecular formula C10H13N4O7PS; molecular weight 364.27 g/mol.
  • CAS number 53-83-8 for unambiguous identification.
  • Purity 90.0% as supplied.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.