Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Trifluoromethyl)-2-pyridinecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. T32701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32701G Supplier TCI America Supplier No. T32701G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorinated pyridine building block with greater than 97.0% purity.

Chemical Identifiers

CAS 131747-42-7
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD02180819
InChI Key OKBHXGBLXDNJJD-UHFFFAOYSA-N
Synonym 6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g
PubChem CID 14761453
IUPAC Name 6-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES OC(=O)C1=NC(=CC=C1)C(F)(F)F

Specifications

Melting Point 157°C
Color White
Quantity 1 g
Formula Weight 191.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Trifluoromethyl)-2-pyridinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.