Learn More
Medchemexpress LLC 6-(Trifluoromethyl)nicotinic acid | 231291-22-8 | 99.9% | C7H4F3NO2 | 10 G
SDP
Catalog No. 5000434231
Encompass_Preferred
6-(Trifluoromethyl)nicotinic acid is a biochemical reagent for life science research. This small heterocyclic molecule, featuring carboxylic acid and trifluoromethyl groups, functions as a versatile additive. In vitro, it can effectively enhance the efficiency and stability of inverted perovskite solar cells.
  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Small heterocyclic molecule with carboxylic acid and trifluoromethyl groups
  • Functions as a multifunctional additive
  • Enhances efficiency and stability of inverted perovskite solar cells (in vitro)
Catalog No. 50-004-34231 Supplier Medchemexpress LLC Supplier No. HY6602910G
6-(Trifluoromethyl)nicotinic acid is a biochemical reagent for life science research. This small heterocyclic molecule, featuring carboxylic acid and trifluoromethyl groups, functions as a versatile additive. In vitro, it can effectively enhance the efficiency and stability of inverted perovskite solar cells.
  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Small heterocyclic molecule with carboxylic acid and trifluoromethyl groups
  • Functions as a multifunctional additive
  • Enhances efficiency and stability of inverted perovskite solar cells (in vitro)