Learn More
Medchemexpress LLC 6-(Trifluoromethyl)nicotinic acid | 231291-22-8 | 99.95% | C7H4F3NO2 | 25 G
SDP
Catalog No. 5000358946
Encompass_Preferred
6-(Trifluoromethyl)nicotinic acid is a biochemical reagent used as a biological material or organic compound for life science research. This small heterocyclic molecule contains carboxylic acid and trifluoromethyl groups, functioning as a multifunctional additive.
  • Used for life science research
  • Contains carboxylic acid and trifluoromethyl groups
  • Functions as a multifunctional additive
  • Enhances efficiency and stability of inverted perovskite solar cells
  • For research use only
Catalog No. 50-003-58946 Supplier Medchemexpress LLC Supplier No. 25GHY66029
6-(Trifluoromethyl)nicotinic acid is a biochemical reagent used as a biological material or organic compound for life science research. This small heterocyclic molecule contains carboxylic acid and trifluoromethyl groups, functioning as a multifunctional additive.
  • Used for life science research
  • Contains carboxylic acid and trifluoromethyl groups
  • Functions as a multifunctional additive
  • Enhances efficiency and stability of inverted perovskite solar cells
  • For research use only