Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%

Catalog No. AAH63138MD
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH63138MD

Supplier: Thermo Scientific Chemicals H63138MD

Only null left
Add to Cart

CAS: 131747-42-7 | C7H4F3NO2 | 191.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131747-42-7
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD02180819
InChI Key OKBHXGBLXDNJJD-UHFFFAOYSA-N
Synonym 6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g
PubChem CID 14761453
IUPAC Name 6-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES OC(=O)C1=NC(=CC=C1)C(F)(F)F

Specifications

Melting Point 154°C to 158°C
Color White
Quantity 250 mg
UN Number UN2811
Formula Weight 191.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 6-(Trifluoromethyl)pyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.