Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, Thermo Scientific™

Catalog No. MAY00261DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00261DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00261DA Supplier Thermo Scientific Chemicals Supplier No. MAY00261DA
Add to Cart

Chemical Identifiers

CAS 959996-58-8
Molecular Formula C6H3ClF3NO2S
Molecular Weight (g/mol) 245.6
InChI Key NSGLNIQAWVNZFB-UHFFFAOYSA-N
Synonym 6-trifluoromethyl pyridine-3-sulfonyl chloride, 6-trifluoromethyl-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride,6-trifluoromethyl, chloro 6-trifluoromethyl 3-pyridyl sulfone, 3-pyridinesulfonyl chloride, 6-trifluoromethyl
PubChem CID 45594311
IUPAC Name 6-(trifluoromethyl)pyridine-3-sulfonyl chloride
SMILES C1=CC(=NC=C1S(=O)(=O)Cl)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 245.6
Percent Purity 97%
Chemical Name or Material 6-(Trifluoromethyl)pyridine-3-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.