Learn More
AdipoGen 6-TRITC (10 mg)
SDP
Supplier:  AdipoGen CDXT0035M010
Encompass_Preferred
6-TRITC (10 mg), AdipoGen Life Sciences. Chemical. CAS: 80724-20-5. Formula: C25H21N3O3S. MW: 443.52. Synthetic. Useful in the fluorescent labeling of proteins. Labeling efficiency with bovine albumin: > 50%.
Catalog No. 50-149-0581