Learn More
Medchemexpress LLC 6-(tritylthio)hexanoic acid | 80441-55-0 | MFCD11226801 | 95.0% | 390.54 g·mol⁻¹ | C25H26O2S | 500 MG
SDP
Catalog No. 5000283181
Encompass_Preferred
6-(Tritylthio)hexanoic acid is a protected thiol linker bearing a tritylthio (Tst) group and a terminal carboxylic acid. The Tst group is removable under acidic conditions to reveal a free thiol. This compound is used as a building block in organic synthesis and for peptide modification and bioconjugation workflows.
Catalog No. 50-002-83181 Supplier Medchemexpress LLC Supplier No. HYW143822500MG
6-(Tritylthio)hexanoic acid is a protected thiol linker bearing a tritylthio (Tst) group and a terminal carboxylic acid. The Tst group is removable under acidic conditions to reveal a free thiol. This compound is used as a building block in organic synthesis and for peptide modification and bioconjugation workflows.