Learn More
Medchemexpress LLC 6-(tritylthio)hexanoic acid | 80441-55-0 | MFCD11226801 | >98.0% | 390.54 g·mol⁻¹ | C25H26O2S | 100 MG
SDP
Catalog No. 5000282866
Encompass_Preferred
6-(Tritylthio)hexanoic acid is a Tst-protected hexanoic acid linker used in organic synthesis and bioconjugation. The tritylthio protecting group can be removed under acidic conditions to reveal a free thiol for thiol-selective conjugations, while the terminal carboxylic acid enables amide bond formation with common coupling reagents.
  • Protecting-group strategy: tritylthio group removable under acidic conditions.
  • Dual functionality: free thiol after deprotection and a terminal carboxylic acid for conjugation.
  • Compatible with standard coupling reagents for reliable amide bond formation.
  • Available in small-scale quantities suitable for synthesis and method development.
  • Solid form suitable for routine laboratory storage and handling.
Catalog No. 50-002-82866 Supplier Medchemexpress LLC Supplier No. HYW143822100MG
6-(Tritylthio)hexanoic acid is a Tst-protected hexanoic acid linker used in organic synthesis and bioconjugation. The tritylthio protecting group can be removed under acidic conditions to reveal a free thiol for thiol-selective conjugations, while the terminal carboxylic acid enables amide bond formation with common coupling reagents.
  • Protecting-group strategy: tritylthio group removable under acidic conditions.
  • Dual functionality: free thiol after deprotection and a terminal carboxylic acid for conjugation.
  • Compatible with standard coupling reagents for reliable amide bond formation.
  • Available in small-scale quantities suitable for synthesis and method development.
  • Solid form suitable for routine laboratory storage and handling.