Learn More
Inmark Inc 6 VAIL AMB D PACK
SDP
Catalog No. NC9154710
Encompass
This item has a minimum qty of 4 per supplier requirements.
Shipper; D-Pack; Frozen vials; EXAKT-PAK;; USDOT Regulations; IATA Packing Instruction 650 for diagnostic specimen transporting; 1 ft. EPS insulation
Catalog No. NC9154710 Supplier Inmark Inc Supplier No. 27222
This item has a minimum qty of 4 per supplier requirements.
Shipper; D-Pack; Frozen vials; EXAKT-PAK;; USDOT Regulations; IATA Packing Instruction 650 for diagnostic specimen transporting; 1 ft. EPS insulation