Learn More
Meiji Techno America Meiji Techno America Halogen lamp, 6V 10W, incident and transmitted light of ABE, ABH, ABZH and PBH stands
Catalog No. NC9642020
Encompass_Preferred
Halogen lamp, 6V 10W, incident and transmitted light of ABE, ABH, ABZH and PBH stands
Catalog No. NC9642020 Supplier Meiji Techno America Supplier No. MA570
Halogen lamp, 6V 10W, incident and transmitted light of ABE, ABH, ABZH and PBH stands