Learn More
Bulbman Inc Osram-Sylvania Micro Bulb, 10 Watt, 6 Volt
SDP
Catalog No. NC9413994
Encompass_Preferred
Micro Bulb; Osram-Sylvania; 6V, 10W; Lumens 200; Base G4; Shape T3; Overall length 31mm; Life Hours 100; Color Clear; 1/EA
Catalog No. NC9413994 Supplier Bulbman Inc Supplier No. 103617
Micro Bulb; Osram-Sylvania; 6V, 10W; Lumens 200; Base G4; Shape T3; Overall length 31mm; Life Hours 100; Color Clear; 1/EA