Learn More
Alkali Scientific ASI™ 6-Well Confocal Plate (35m), Clear, Adhesion Type, Sterile, 4/Cs, PS/Flux, Treated (Positive-Charged/ Surface Coated), 50/Cs, 22 x 22 x 0.13mm, Hole 1.33mm
SDP
Catalog No. 503028861
Encompass_Preferred
ASI™ confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products: Glass: naturally charged, good growth surface for adherent cellsFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy ▪ Bottom materials: Glass/FLux (no surface treatment for glass bottom)▪ Dish color: Clear▪ Dish size: 35 Ø▪ Plate size: 6well Plate▪ Hole size: 13 Ø / 20 Ø / 25Ø▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free
Catalog No. 50-302-8861 Supplier Alkali Scientific Supplier No. TCP23
ASI™ confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products: Glass: naturally charged, good growth surface for adherent cellsFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy ▪ Bottom materials: Glass/FLux (no surface treatment for glass bottom)▪ Dish color: Clear▪ Dish size: 35 Ø▪ Plate size: 6well Plate▪ Hole size: 13 Ø / 20 Ø / 25Ø▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free